Merck
  • A novel Fe(II)/citrate/UV/peroxymonosulfate process for micropollutant degradation: Optimization by response surface methodology and effects of water matrices.
A novel Fe(II)/citrate/UV/peroxymonosulfate process for micropollutant degradation: Optimization by response surface methodology and effects of water matrices.
Chemosphere (2017-06-15)
Li Ling, Dapeng Zhang, Jingyun Fang, Chihhao Fan, Chii Shang
ZUSAMMENFASSUNG
This paper applied the response surface methodology (RSM) to optimizing a novel Fe(II)/citrate/UV/PMS process in the degradation of a model micropollutant, carbamazepine (CBZ), a persistent emerging contaminant frequently detected in surface water and groundwater. The experimental conditions in terms of two responses, CBZ removal efficiency (Y1) and cost per unit CBZ removal (Y2), were optimized by the central composite design (CCD) in RSM. Modeling data exhibited that the optimum condition resulting in the lowest Y2 while achieving >70% of Y1 was at a UV dose of 265.5 mJ/cm
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phenazin-Methosulfat