Merck
Acute Increases in Protein O-GlcNAcylation Dampen Epileptiform Activity in Hippocampus.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2017-08-02)
Luke T Stewart, Anas U Khan, Kai Wang, Diana Pizarro, Sandipan Pati, Susan C Buckingham, Michelle L Olsen, John C Chatham, Lori L McMahon
ZUSAMMENFASSUNG
O-GlcNAcylation is a ubiquitous and dynamic post-translational modification involving the O-linkage of β-
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pentylentetrazol
Sigma-Aldrich
Picrotoxin, powder