Merck
  • Defective N-sulfation of heparan sulfate proteoglycans limits PDGF-BB binding and pericyte recruitment in vascular development.
Defective N-sulfation of heparan sulfate proteoglycans limits PDGF-BB binding and pericyte recruitment in vascular development.
Genes & development (2007-02-10)
Alexandra Abramsson, Sindhulakshmi Kurup, Marta Busse, Shuhei Yamada, Per Lindblom, Edith Schallmeiner, Denise Stenzel, Dominique Sauvaget, Johan Ledin, Maria Ringvall, Ulf Landegren, Lena Kjellén, Göran Bondjers, Jin-ping Li, Ulf Lindahl, Dorothe Spillmann, Christer Betsholtz, Holger Gerhardt
ZUSAMMENFASSUNG
During vascular development, endothelial platelet-derived growth factor B (PDGF-B) is critical for pericyte recruitment. Deletion of the conserved C-terminal heparin-binding motif impairs PDGF-BB retention and pericyte recruitment in vivo, suggesting a potential role for heparan sulfate (HS) in PDGF-BB function during vascular development. We studied the participation of HS chains in pericyte recruitment using two mouse models with altered HS biosynthesis. Reduction of N-sulfation due to deficiency in N-deacetylase/N-sulfotransferase-1 attenuated PDGF-BB binding in vitro, and led to pericyte detachment and delayed pericyte migration in vivo. Reduced N-sulfation also impaired PDGF-BB signaling and directed cell migration, but not proliferation. In contrast, HS from glucuronyl C5-epimerase mutants, which is extensively N- and 6-O-sulfated, but lacks 2-O-sulfated L-iduronic acid residues, retained PDGF-BB in vitro, and pericyte recruitment in vivo was only transiently delayed. These observations were supported by in vitro characterization of the structural features in HS important for PDGF-BB binding. We conclude that pericyte recruitment requires HS with sufficiently extended and appropriately spaced N-sulfated domains to retain PDGF-BB and activate PDGF receptor beta (PDGFRbeta) signaling, whereas the detailed sequence of monosaccharide and sulfate residues does not appear to be important for this interaction.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Platelet-Derived Growth Factor-BB from rat, PDGF-BB, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture