Merck
  • Nickel(0) nanoparticles supported on bare or coated cobalt ferrite as highly active, magnetically isolable and reusable catalyst for hydrolytic dehydrogenation of ammonia borane.
Nickel(0) nanoparticles supported on bare or coated cobalt ferrite as highly active, magnetically isolable and reusable catalyst for hydrolytic dehydrogenation of ammonia borane.
Journal of colloid and interface science (2017-08-28)
Joydev Manna, Serdar Akbayrak, Saim Özkar
ZUSAMMENFASSUNG
Nickel(0) nanoparticles supported on cobalt ferrite (Ni
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Boranammoniak-Komplex, 95%
Sigma-Aldrich
Nickel(II)-chlorid Hexahydrat, 99.9% trace metals basis