Merck
ZNF516 suppresses EGFR by targeting the CtBP/LSD1/CoREST complex to chromatin.
Nature communications (2017-09-28)
Lifang Li, Xinhua Liu, Lin He, Jianguo Yang, Fei Pei, Wanjin Li, Shumeng Liu, Zhe Chen, Guojia Xie, Bosen Xu, Xia Ting, Zihan Zhang, Tong Jin, Xujun Liu, Wenting Zhang, Shuai Yuan, Ziran Yang, Chongyang Wu, Yu Zhang, Xiaohan Yang, Xia Yi, Jing Liang, Yongfeng Shang, Luyang Sun
ZUSAMMENFASSUNG
EGFR is required for animal development, and dysregulation of EGFR is critically implicated in malignant transformation. However, the molecular mechanism underlying the regulation of EGFR expression remains poorly explored. Here we report that the zinc-finger protein ZNF516 is a transcription repressor. ZNF516 is physically associated with the CtBP/LSD1/CoREST complex and transcriptionally represses a cohort of genes including EGFR that are critically involved in cell proliferation and motility. We demonstrate that the ZNF516-CtBP/LSD1/CoREST complex inhibits the proliferation and invasion of breast cancer cells in vitro and suppresses breast cancer growth and metastasis in vivo. Significantly, low expression of ZNF516 is positively associated with advanced pathological staging and poor survival of breast carcinomas. Our data indicate that ZNF516 is a transcription repressor and a potential suppressor of EGFR, adding to the understanding of EGFR-related breast carcinogenesis and supporting the pursuit of ZNF516 as a potential therapeutic target for breast cancer. EGFR is a well-known oncogene; however, the mechanisms regulating its expression are still unclear. Here, analysing genome-wide chromatin associations, the authors show that in breast cancer cells ZNF516 represses EGFR transcription through the interaction with the CtBP/LSD1/CoREST complex.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
ANTI-FLAG® M2-Affinitätsgel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Trichostatin A, Ready Made Solution, 5 mM in DMSO, from Streptomyces sp.
Sigma-Aldrich
Monoclonal Anti-Fibronectin antibody produced in mouse, clone FN-15, ascites fluid