- Direct rate assessment of laccase catalysed radical formation in lignin by electron paramagnetic resonance spectroscopy.
Direct rate assessment of laccase catalysed radical formation in lignin by electron paramagnetic resonance spectroscopy.
Enzyme and microbial technology (2017-09-02)
Line Munk, Mogens Larsen Andersen, Anne S Meyer
PMID28859815
ZUSAMMENFASSUNG
Laccases (EC 1.10.3.2) catalyse removal of an electron and a proton from phenolic hydroxyl groups, including phenolic hydroxyls in lignins, to form phenoxy radicals during reduction of O