Merck
  • Automated T2-mapping of the Menisci From Magnetic Resonance Images in Patients with Acute Knee Injury.
Automated T2-mapping of the Menisci From Magnetic Resonance Images in Patients with Acute Knee Injury.
Academic radiology (2017-05-30)
Anthony Paproki, Craig Engstrom, Mark Strudwick, Katharine J Wilson, Rachel K Surowiec, Charles Ho, Stuart Crozier, Jurgen Fripp
ZUSAMMENFASSUNG
This study aimed to evaluate the accuracy of an automated method for segmentation and T2 mapping of the medial meniscus (MM) and lateral meniscus (LM) in clinical magnetic resonance images from patients with acute knee injury. Eighty patients scheduled for surgery of an anterior cruciate ligament or meniscal injury underwent magnetic resonance imaging of the knee (multiplanar two-dimensional [2D] turbo spin echo [TSE] or three-dimensional [3D]-TSE examinations, T2 mapping). Each meniscus was automatically segmented from the 2D-TSE (composite volume) or 3D-TSE images, auto-partitioned into anterior, mid, and posterior regions, and co-registered onto the T2 maps. The Dice similarity index (spatial overlap) was calculated between automated and manual segmentations of 2D-TSE (15 patients), 3D-TSE (16 patients), and corresponding T2 maps (31 patients). Pearson and intraclass correlation coefficients (ICC) were calculated between automated and manual T2 values. T2 values were compared (Wilcoxon rank sum tests) between torn and non-torn menisci for the subset of patients with both manual and automated segmentations to compare statistical outcomes of both methods. The Dice similarity index values for the 2D-TSE, 3D-TSE, and T2 map volumes, respectively, were 76.4%, 84.3%, and 75.2% for the MM and 76.4%, 85.1%, and 76.1% for the LM. There were strong correlations between automated and manual T2 values (r The present automated method offers a promising alternative to manual T2 mapping analyses of the menisci and a considerable advance for integration into clinical workflows.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3,3′-Diiod-L-thyronin (T2), 98% (CP)