Merck
The Exometabolome of Two Model Strains of the
Frontiers in microbiology (2017-10-28)
Gerrit Wienhausen, Beatriz E Noriega-Ortega, Jutta Niggemann, Thorsten Dittmar, Meinhard Simon
ZUSAMMENFASSUNG
Recent studies applying Fourier transform ion cyclotron resonance mass spectrometry (FT-ICR-MS) showed that the exometabolome of marine bacteria is composed of a surprisingly high molecular diversity. To shed more light on how this diversity is generated we examined the exometabolome of two model strains of the