Merck
  • Cytoplasmic neuropilin 2 is associated with metastasis and a poor prognosis in early tongue cancer patients.
Cytoplasmic neuropilin 2 is associated with metastasis and a poor prognosis in early tongue cancer patients.
International journal of oral and maxillofacial surgery (2017-06-13)
H S Ong, S Gokavarapu, Q Xu, Z Tian, J Li, T Ji, C P Zhang
ZUSAMMENFASSUNG
Neuropilin 2 (Nrp2) plays an important role in regulating lymphangiogenesis. Nrp2 expression in early tongue cancer was investigated to predict lymph node metastasis and the long-term prognosis. The relationships between clinicopathological variables of cT1-T2N0 tongue squamous cell carcinoma (SCC) and overexpression of Nrp2, vascular endothelial growth factor C (VEGFC), vascular endothelial growth factor receptor 3 (VEGFR3), and semaphorin 3F (Sema3F) were analyzed. Expression levels were compared using oral SCC cell lines. The Nrp2 gene was silenced to determine the impact of Nrp2. Cytoplasmic Nrp2 overexpression predicted regional metastasis with sensitivity and specificity of 90.3% and 42.1%, respectively. Cytoplasmic Nrp2 overexpression (P<0.001) and VEGFC overexpression (P=0.006) were significantly related to regional metastasis (Student t-test). However, only cytoplasmic Nrp2 overexpression was an independent prognostic factor for both disease-free survival (DFS; P=0.008) and overall survival (OS; P=0.016) (Cox regression); the risk of recurrence was 12-times higher (P=0.015) and risk of mortality was 8-times higher (P=0.016). Co-localization of Nrp2 and VEGFC was greater within the cytoplasm of aggressive cell lines (HN12 and RCa-T). Nrp2 plays a role in tumourigenesis; VEGFC supplementation cannot rescue the biological function of Nrp2 in Nrp2-depleted cell lines. Cytoplasmic Nrp2 overexpression is associated with decreased OS and DFS. Cytoplasmic Nrp2 overexpression may be a reliable diagnostic and prognostic marker for early tongue SCC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin-Fragment 26-33 Amid, ≥97% (HPLC), powder