Merck
Mutation in human
Proceedings of the National Academy of Sciences of the United States of America (2017-09-07)
Yvette Y Yien, Sarah Ducamp, Lisa N van der Vorm, Julia R Kardon, Hana Manceau, Caroline Kannengiesser, Hector A Bergonia, Martin D Kafina, Zoubida Karim, Laurent Gouya, Tania A Baker, Hervé Puy, John D Phillips, Gaël Nicolas, Barry H Paw
ZUSAMMENFASSUNG
Loss-of-function mutations in genes for heme biosynthetic enzymes can give rise to congenital porphyrias, eight forms of which have been described. The genetic penetrance of the porphyrias is clinically variable, underscoring the role of additional causative, contributing, and modifier genes. We previously discovered that the mitochondrial AAA+ unfoldase ClpX promotes heme biosynthesis by activation of δ-aminolevulinate synthase (ALAS), which catalyzes the first step of heme synthesis. CLPX has also been reported to mediate heme-induced turnover of ALAS. Here we report a dominant mutation in the ATPase active site of human CLPX, p.Gly298Asp, that results in pathological accumulation of the heme biosynthesis intermediate protoporphyrin IX (PPIX). Amassing of PPIX in erythroid cells promotes erythropoietic protoporphyria (EPP) in the affected family. The mutation in