- Prostaglandin F
Prostaglandin F
Reproduction, fertility, and development (2017-03-17)
Shuangyi Zhang, Bo Liu, Long Gao, Wei Mao, Changqi Fu, Duritahala Duritahala, Nan Zhang, Ying Zhang, Yuan Shen, Jinshan Cao
PMID28298246
ZUSAMMENFASSUNG
The endometrium of domestic animals undergoes regular periods of regeneration and degeneration and exhibits a remarkable capacity for self-repair during the oestrous cycle. The endometrial growth pattern is also observed during in the implantation period and early pregnancy, but the mechanism underlying endometrial growth in these processes remains unclear. A positive correlation between endometrial growth in these processes and prostaglandin (PG) F