Merck
  • Micropropagation and Subsequent Enrichment of Carotenoids, Fatty Acids, and Tocopherol Contents in
Micropropagation and Subsequent Enrichment of Carotenoids, Fatty Acids, and Tocopherol Contents in
Frontiers in chemistry (2017-10-25)
Han Yong Park, Ramesh Kumar Saini, Judy Gopal, Young-Soo Keum, Doo Hwan Kim, Onew Lee, Iyyakkannu Sivanesan
ZUSAMMENFASSUNG
A promising micropropagation protocol has been systematically established and demonstrated for the enhanced production of carotenoids, tocopherol and fatty acids in shoot tissues of
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
LDN-57444, ≥98% (HPLC)