Merck
NLRP3 Phosphorylation Is an Essential Priming Event for Inflammasome Activation.
Molecular cell (2017-09-26)
Nan Song, Zhao-Shan Liu, Wen Xue, Zhao-Fang Bai, Qian-Yi Wang, Jiang Dai, Xin Liu, Yi-Jiao Huang, Hong Cai, Xiao-Yan Zhan, Qiu-Ying Han, Hongxia Wang, Yuan Chen, Hui-Yan Li, Ai-Ling Li, Xue-Min Zhang, Tao Zhou, Tao Li
ZUSAMMENFASSUNG
Many infections and stress signals can rapidly activate the NLRP3 inflammasome to elicit robust inflammatory responses. This activation requires a priming step, which is thought to be mainly for upregulating NLRP3 transcription. However, recent studies report that the NLRP3 inflammasome can be activated independently of transcription, suggesting that the priming process has unknown essential regulatory steps. Here, we report that JNK1-mediated NLRP3 phosphorylation at S194 is a critical priming event and is essential for NLRP3 inflammasome activation. We show that NLRP3 inflammasome activation is disrupted in NLRP3-S194A knockin mice. JNK1-mediated NLRP3 S194 phosphorylation is critical for NLRP3 deubiquitination and facilitates its self-association and the subsequent inflammasome assembly. Importantly, we demonstrate that blocking S194 phosphorylation prevents NLRP3 inflammasome activation in cryopyrin-associated periodic syndromes (CAPS). Thus, our study reveals a key priming molecular event that is a prerequisite for NLRP3 inflammasome activation. Inhibiting NLRP3 phosphorylation could be an effective treatment for NLRP3-related diseases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Roche
cOmplete Proteasehemmer-Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Monoklonaler Anti-α-Tubulin-Antikörper in Maus hergestellte Antikörper, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Resiquimod, ≥98% (HPLC)
Sigma-Aldrich
IRAK-1/4 Inhibitor I, ≥98% (HPLC), solid