Merck
  • Production of bFGF monoclonal antibody and its inhibition of metastasis in Lewis lung carcinoma.
Production of bFGF monoclonal antibody and its inhibition of metastasis in Lewis lung carcinoma.
Molecular medicine reports (2017-08-03)
Yanqing Yang, Zhenming Luo, Yiyang Qin, Yu Zhou, Longcai Gong, Jianfang Huang, Hong Wang
ZUSAMMENFASSUNG
Basic fibroblast growth factor (bFGF) and fibroblast growth factor receptor 1 (FGFR1) are associated with drug resistance in lung cancer. In the present study, mouse monoclonal antibodies (mAb) against human bFGF, targeting the binding site of bFGF with FGFR1 were produced, and the antitumor activity and inhibition of metastasis was studied in Lewis lung carcinoma (LLC). A total of four hybridoma cell strains that stably secreted bFGF mAb were obtained. mAbE12 was selected as the most effective for use in the following studies, with a relative affinity constant of 5.66x108 l/mol. mAbE12 was demonstrated to inhibit cell proliferation and tumor growth in vitro and in vivo. Furthermore, mAbE12 blocked migration and metastasis of LLC cells in vitro and in vivo. This occurred due to a mAbE12‑induced upregulation of E‑cadherin expression through the protein kinase B‑glycogen synthase kinase 3 β‑Snail pathway. These results suggested that mAbE12 may be a potential antibody for the treatment of lung cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin-Fragment 26-33 Amid, ≥97% (HPLC), powder