- The pharmacological regulation of cellular mitophagy.
The pharmacological regulation of cellular mitophagy.
Nature chemical biology (2017-01-20)
Nikolaos D Georgakopoulos, Geoff Wells, Michelangelo Campanella
PMID28103219
ZUSAMMENFASSUNG
Small molecules are pharmacological tools of considerable value for dissecting complex biological processes and identifying potential therapeutic interventions. Recently, the cellular quality-control process of mitophagy has attracted considerable research interest; however, the limited availability of suitable chemical probes has restricted our understanding of the molecular mechanisms involved. Current approaches to initiate mitophagy include acute dissipation of the mitochondrial membrane potential (ΔΨ