Merck
  • Design, Synthesis, and Evaluation of Triazole Derivatives That Induce Nrf2 Dependent Gene Products and Inhibit the Keap1-Nrf2 Protein-Protein Interaction.
Design, Synthesis, and Evaluation of Triazole Derivatives That Induce Nrf2 Dependent Gene Products and Inhibit the Keap1-Nrf2 Protein-Protein Interaction.
Journal of medicinal chemistry (2015-09-09)
Hélène C Bertrand, Marjolein Schaap, Liam Baird, Nikolaos D Georgakopoulos, Adrian Fowkes, Clarisse Thiollier, Hiroko Kachi, Albena T Dinkova-Kostova, Geoff Wells
ZUSAMMENFASSUNG
The transcription factor Nrf2 regulates the expression of a large network of cytoprotective and metabolic enzymes and proteins. Compounds that directly and reversibly inhibit the interaction between Nrf2 and its main negative regulator Keap1 are potential pharmacological agents for a range of disease types including neurodegenerative conditions and cancer. We describe the development of a series of 1,4-diphenyl-1,2,3-triazole compounds that inhibit the Nrf2-Keap1 protein-protein interaction (PPI) in vitro and in live cells and up-regulate the expression of Nrf2-dependent gene products.