- Effects of H
Effects of H
Physical chemistry chemical physics : PCCP (2017-10-05)
Yoshikata Koga, Kumiko Miki, Keiko Nishikawa
PMID28975179
ZUSAMMENFASSUNG
We applied what we call the "1-propanol (1P) probing methodology" on the effects of H
MATERIALIEN