Merck
  • Integrated analysis of coding genes and non-coding RNAs during hair follicle cycle of cashmere goat (Capra hircus).
Integrated analysis of coding genes and non-coding RNAs during hair follicle cycle of cashmere goat (Capra hircus).
BMC genomics (2017-10-13)
Shanhe Wang, Wei Ge, Zhixin Luo, Yang Guo, Beilei Jiao, Lei Qu, Zhiying Zhang, Xin Wang
ZUSAMMENFASSUNG
Cashmere growth is a seasonal and cyclic phenomenon under the control of photoperiod and multiple stimulatory and inhibitory signals. Beyond relevant coding genes, microRNA (miRNA) and long non coding RNA (lncRNA) play an indispensable role in hair follicle (HF) development and skin homeostasis. Furthermore, the influence of lncRNA upon miRNA function is also rapidly emerging. However, little is known about miRNAs, lncRNAs and their functions as well as their interactions on cashmere development and cycling. Here, based on lncRNA and miRNA high-throughput sequencing and bioinformatics analysis, we have identified 1108 lncRNAs and 541 miRNAs in cashmere goat skin during anagen and telogen. Compared with telogen, 1388 coding genes, 41 lncRNAs and 15 miRNAs were upregulated, while 1104 coding genes, 157 lncRNAs and 8 miRNAs were downregulated in anagen (adjusted P-value ≤0.05 and relative fold-change ≥2). Subsequently, we investigated the impact of lncRNAs on their target genes in cis and trans, indicating that these lncRNAs are functionally conserved during HF development and cycling. Furthermore, miRNA-mRNA and miRNA-lncRNA interaction were identified through the bioinformatics algorithm miRanda, then the ceRNA networks, miR-221-5p-lnc_000679-WNT3, miR-34a-lnc_000181-GATA3 and miR-214-3p-lnc_000344-SMAD3, were constructed under defined rules, to illustrate their roles in cashmere goat HF biology. The present study provides a resource for lncRNA, miRNA and mRNA studies in cashmere cycling and development. We also demonstrate potential ceRNA regulatory networks in cashmere goat HF cycling for the first time. It expands our knowledge about lncRNA and miRNA biology as well as contributes to the annotation of the goat genome.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Octanoyl-N-Methylglucamin, ≥97% (GC)