Merck
  • Fulvic acid anchored layered double hydroxides: A multifunctional composite adsorbent for the removal of anionic dye and toxic metal.
Fulvic acid anchored layered double hydroxides: A multifunctional composite adsorbent for the removal of anionic dye and toxic metal.
Journal of hazardous materials (2017-09-22)
Lei Li, Guangxia Qi, Bangda Wang, Dongbei Yue, Yi Wang, Tsutomu Sato
ZUSAMMENFASSUNG
A novel multifunctional composite adsorbent which possesses the ability for anion exchange and toxic metal complexation has been synthesized by the hybridization of layered double hydroxides (LDH) and fulvic acid (FA) in this study. The results show that FA with lots of functional groups can be effectively and stably anchored on the surface of LDH through coagulation process without occupying the interlayer of LDH. Therefore, the anion exchange ability remains and the adsorption capacity of Orange II can reach 1.9mmol/g, which is almost as much as stoichiometric anion exchange capacity of pure LDH. Moreover, the composite adsorbent's adsorption capacity of Cu