- Selective Colorimetric Detection of Nitrite in Water using Chitosan Stabilized Gold Nanoparticles Decorated Reduced Graphene oxide.
Selective Colorimetric Detection of Nitrite in Water using Chitosan Stabilized Gold Nanoparticles Decorated Reduced Graphene oxide.
Scientific reports (2017-10-29)
Baishnisha Amanulla, Selvakumar Palanisamy, Shen-Ming Chen, Te-Wei Chiu, Vijayalakshmi Velusamy, James M Hall, Tse-Wei Chen, Sayee Kannan Ramaraj
PMID29079840
ZUSAMMENFASSUNG
Excess nitrite (NO