Merck
  • Inclusion complex from cyclodextrin-grafted hyaluronic acid and pseudo protein as biodegradable nano-delivery vehicle for gambogic acid.
Inclusion complex from cyclodextrin-grafted hyaluronic acid and pseudo protein as biodegradable nano-delivery vehicle for gambogic acid.
Acta biomaterialia (2017-09-02)
Ying Ji, Shuo Shan, Mingyu He, Chih-Chang Chu
ZUSAMMENFASSUNG
β-Cyclodextrin can form inclusion complex with a series of guest molecules including phenyl moieties, and has gained considerable popularity in the study of supramolecular nanostructure. In this study, a biodegradable nanocomplex (HA(CD)-4Phe4 nanocomplex) was developed from β-cyclodextrin grafted hyaluronic acid (HA) and phenylalanine based poly(ester amide). The phenylalanine based poly(ester amide) is a biodegradable pseudo protein which provides the encapsulation capacity for gambogic acid (GA), a naturally-derived chemotherapeutic which has been effectively employed to treat multidrug resistant tumor. The therapeutic potency of free GA is limited due to its poor solubility in water and the lack of tumor-selective toxicity. The nanocomplex carrier enhanced the solubility and availability of GA in aqueous media, and the HA component enabled the targeted delivery to tumor cells with overexpression of CD44 receptors. In the presence of hyaluronidase, the release of GA from the nanocomplex was significantly accelerated, due to the enzymatic biodegradation of the carrier. Compared to free GA, GA-loaded nanocomplex exhibited improved cytotoxicity in MDA-MB-435/MDR multidrug resistant melanoma cells, and induced enhanced level of apoptosis and mitochondrial depolarization, at low concentration of GA (1-2µM). The nanocomplex enhanced the therapeutic potency of GA, especially when diluted in physiological environment. In addition, suppressed matrix metalloproteinase activity was also detected in MDA-MB-435/MDR cells treated by GA-loaded nanocomplex, which demonstrated its potency in the inhibition of tumor metastasis. The in vitro data suggested that HA(CD)-4Phe4 nanocomplex could provide a promising alternative in the treatment of multidrug resistant tumor cells. Gambogic acid (GA), naturally derived from genus Garcinia trees, exhibited significant cytotoxic activity against multiple types of tumors with resistance to traditional chemotherapeutics. Unfortunately, the poor solubility of GA in conventional pharmaceutical solvents and non-targeted distribution in normal tissues greatly limited its therapeutic potency. To overcome the challenges, we develop a nanoplatform from the supramolecular assembly of β-cyclodextrin grafted hyaluronic acid (HA) and phenylalanine based pseudo protein. The pseudo protein in the nanocomplex provided the hydrophobic interaction and loading capacity for GA, while the HA component targeted the overexpressed CD44 receptor and improved the selective endocytosis in multidrug resistant melanoma cells. The supramolecular nanocomplex provide a promising platform for the delivery of hydrophobic chemotherapeutics to improve the bioavailability and efficiency.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-Adamantancarbonsäure, 99%
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Sigma-Aldrich
Bernsteinsäuredihydrazid, 96%