Merck
  • Mycotoxins, trace elements, and phthalates in marketed rice of different origin and exposure assessment.
Mycotoxins, trace elements, and phthalates in marketed rice of different origin and exposure assessment.
Food additives & contaminants. Part B, Surveillance (2017-06-18)
Biljana D Škrbić, Yaqin Ji, Jelena R Živančev, Grigorije G Jovanović, Zhao Jie
ZUSAMMENFASSUNG
The aim of this study was to determine levels of 11 mycotoxins, 10 trace elements, and 6 phthalates in rice samples from Serbian and Chinese market. Mycotoxins were not detected in any of the analysed rice samples. Results revealed similar median levels for following elements: Mn, 17.5 and 15.7 mg kg
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dess-Martin-Periodinan, 97%
Supelco
T2-Toxin -Lösung, 100 μg/mL in acetonitrile, analytical standard
Supelco
Fumonisin B1 -Lösung, ~50 μg/mL in acetonitrile: water, analytical standard
Supelco
Fumonisin B2 -Lösung, ~50 μg/mL in acetonitrile: water (50:50), analytical standard
Supelco
HT-2 Toxin -Lösung, 100 μg/mL in acetonitrile, analytical standard