- Ion-macromolecule interactions studied with model polyurethanes.
Ion-macromolecule interactions studied with model polyurethanes.
Journal of colloid and interface science (2017-09-12)
Borja Fernández-d'Arlas, Miguel Ángel Huertos, Alejandro J Müller
PMID28892743
ZUSAMMENFASSUNG
The solubility and self-assembly of macromolecules in solution can be tuned by the presence of different salts. Natural proteins have been long manipulated with the aid of salts, and natural silk is processed in the gland tip across a gradient of different salts which modifies its solubility. Hence, the comprehensive understanding of the role of ion-macromolecule interactions should pave the way towards a biomimetic processing of macromolecules. A model polyurethane catiomer (PU The PU