Merck
Novel mechanism of regulation of the 5-lipoxygenase/leukotriene B
Scientific reports (2017-10-13)
Shigeyasu Tsuda, Masakazu Shinohara, Toshihiko Oshita, Manabu Nagao, Nobuaki Tanaka, Takeshige Mori, Tetsuya Hara, Yasuhiro Irino, Ryuji Toh, Tatsuro Ishida, Ken-Ichi Hirata
ZUSAMMENFASSUNG
High-density lipoprotein (HDL) interacts with various cells, particularly macrophages, in functional cell-HDL interactions. Here, we found that HDL protein quality and lipid quality play critical roles in HDL functions. HDL fractions from healthy volunteers (HDL
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
BLT-1, ≥98% (HPLC)