Merck
  • Peroxisomal CuAOζ and its product H2O2 regulate the distribution of auxin and IBA-dependent lateral root development in Arabidopsis.
Peroxisomal CuAOζ and its product H2O2 regulate the distribution of auxin and IBA-dependent lateral root development in Arabidopsis.
Journal of experimental botany (2017-10-11)
Yana Qu, Qing Wang, Jinhe Guo, Peipei Wang, Ping Song, Qianru Jia, Xinxin Zhang, Jörg Kudla, Wenhua Zhang, Qun Zhang
ZUSAMMENFASSUNG
Root system architecture depends on endogenous and environmental signals, including polar transport of the phytohormone auxin, reactive oxygen species (ROS), nutrient availability, and stresses. In our study, we describe a novel Arabidopsis thaliana peroxisome-localized copper amine oxidase ζ (CuAOζ), which is highly expressed in cortical cells, and the ROS derived from CuAOζ are essential for lateral root (LR) development. Loss of CuAOζ results in retarded auxin-induced ROS generation, PINFORMED2 (PIN2)-mediated auxin transport, and LR development in response to added indole-3-butyric acid. Auxins enhance CuAOζ protein levels and their cellular translocation toward the plasma membrane in the cortex. CuAOζ interacts physically with PEROXINS5 via an N-terminal signal tag, Ser-Lys-Leu, and is transported into the peroxisome upon this interaction, which is required for the functions of CuAOζ in the auxin response. Together, our results suggest a peroxisomal ROS-based auxin signaling pathway involving spatiotemporal-dependent CuAOζ functional regulation of PIN2 homeostasis, auxin distribution, and LR development.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Indole-3-butyric acid potassium salt, suitable for plant cell culture, BioReagent