Merck
  • Molecular analysis of oxalate-induced endoplasmic reticulum stress mediated apoptosis in the pathogenesis of kidney stone disease.
Molecular analysis of oxalate-induced endoplasmic reticulum stress mediated apoptosis in the pathogenesis of kidney stone disease.
Journal of physiology and biochemistry (2017-09-07)
Albert Abhishek, Shaly Benita, Monika Kumari, Divya Ganesan, Eldho Paul, Ponnusamy Sasikumar, Ayyavu Mahesh, Subramani Yuvaraj, Tharmarajan Ramprasath, Govindan Sadasivam Selvam
ZUSAMMENFASSUNG
Oxalate, a non-essential end product of metabolism, causes hyperoxaluria and eventually calcium oxalate (CaOx) stone disease. Kidney cells exposed to oxalate stress results in generation of reactive oxygen species (ROS) and progression of stone formation. Perturbations in endoplasmic reticulum (ER) result in accumulation of misfolded proteins and Ca
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thioflavin T, used as stain for amyloid
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)