Merck
  • Multi-response modeling of reaction-diffusion to explain alpha-galactoside behavior during the soaking-cooking process in cowpea.
Multi-response modeling of reaction-diffusion to explain alpha-galactoside behavior during the soaking-cooking process in cowpea.
Food chemistry (2017-10-19)
Fanny Coffigniez, Aurélien Briffaz, Christian Mestres, Pascaline Alter, Noel Durand, Philippe Bohuon
ZUSAMMENFASSUNG
A modelling approach was developed to better understand the behavior of the flatulence-causing oligosaccharides in cowpea seeds during isothermal water soaking-cooking process. Concentrations of verbascose, stachyose and raffinose were measured both in the seed and in the soaking water during the process (T=30, 60 and 95°C). A reaction-diffusion model was built for the three considered alpha-galactosides both in the seed and in the soaking water, together with a model of water transport in the seed. The model reproduced coupled reaction-diffusion of alpha-galactosides during the soaking-cooking process with a good fit. Produced, diffused and degraded alpha-galactoside fractions were identified by performing a mass balance. During soaking at 30°C, degradation predominated (maximum found for raffinose degradation rate constant of 3.22×10
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natronlauge, 50% in H2O