Merck
A homodimer interface without base pairs in an RNA mimic of red fluorescent protein.
Nature chemical biology (2017-09-26)
Katherine Deigan Warner, Ljiljana Sjekloća, Wenjiao Song, Grigory S Filonov, Samie R Jaffrey, Adrian R Ferré-D'Amaré
ZUSAMMENFASSUNG
Corn, a 28-nucleotide RNA, increases yellow fluorescence of its cognate ligand 3,5-difluoro-4-hydroxybenzylidene-imidazolinone-2-oxime (DFHO) by >400-fold. Corn was selected in vitro to overcome limitations of other fluorogenic RNAs, particularly rapid photobleaching. We now report the Corn-DFHO co-crystal structure, discovering that the functional species is a quasisymmetric homodimer. Unusually, the dimer interface, in which six unpaired adenosines break overall two-fold symmetry, lacks any intermolecular base pairs. The homodimer encapsulates one DFHO at its interprotomer interface, sandwiching it with a G-quadruplex from each protomer. Corn and the green-fluorescent Spinach RNA are structurally unrelated. Their convergent use of G-quadruplexes underscores the usefulness of this motif for RNA-induced small-molecule fluorescence. The asymmetric dimer interface of Corn could provide a basis for the development of mutants that only fluoresce as heterodimers. Such variants would be analogous to Split GFP, and may be useful for analyzing RNA co-expression or association, or for designing self-assembling RNA nanostructures.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thioflavin T, used as stain for amyloid