Merck
Suppression of ciliary movements by a hypertonic stress in the newt olfactory receptor neuron.
American journal of physiology. Cell physiology (2017-07-08)
Yoshihiko Wakazono, Takashi Sakurai, Susumu Terakawa
ZUSAMMENFASSUNG
Olfactory receptor neurons isolated from the newt maintain a high activity of the ciliary beat. A cilium of neuron is so unique that only little is known about regulatory factors for its beat frequency. We examined the olfactory receptor neuron immersed in various extracellular media under the video-enhanced differential interference contrast microscope. The activation of voltage-gated Ca
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triton X-100, laboratory grade