Merck
  • Proteomic Analysis of Thiol Modifications and Assessment of Structural Changes in Hemoglobin Induced by the Aniline Metabolites N-Phenylhydroxylamine and Nitrosobenzene.
Proteomic Analysis of Thiol Modifications and Assessment of Structural Changes in Hemoglobin Induced by the Aniline Metabolites N-Phenylhydroxylamine and Nitrosobenzene.
Scientific reports (2017-11-03)
Carolina Möller, W Clay Davis, Vanessa R Thompson, Frank Marí, Anthony P DeCaprio
ZUSAMMENFASSUNG
MS-based proteomic analysis was combined with in silico quantum mechanical calculations to improve understanding of protein adduction by N-phenylhydroxylamine (PhNHOH) and nitrosobenzene (NOB), metabolic products of aniline. In vitro adduction of model peptides containing nucleophilic sidechains (Cys, His, and Lys) and selected proteins (bovine and human hemoglobin and β-lactoglobulin-A) were characterized. Peptide studies identified the Cys thiolate as the most reactive nucleophile for these metabolites, a result consistent with in silico calculations of reactivity parameters. For PhNHOH, sulfinamides were identified as the primary adduction products, which were stable following tryptic digestion. Conversely, reactions with NOB yielded an additional oxidized adduct, the sulfonamide. In vitro exposure of human whole blood to PhNHOH and NOB demonstrated that only sulfinamides were formed. In addition to previously reported adduction of β
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nitrosobenzol, ≥97%