Merck
  • The Anti-Inflammatory Effect of Ripasudil (K-115), a Rho Kinase (ROCK) Inhibitor, on Endotoxin-Induced Uveitis in Rats.
The Anti-Inflammatory Effect of Ripasudil (K-115), a Rho Kinase (ROCK) Inhibitor, on Endotoxin-Induced Uveitis in Rats.
Investigative ophthalmology & visual science (2017-10-31)
Takatoshi Uchida, Megumi Honjo, Reiko Yamagishi, Makoto Aihara
ZUSAMMENFASSUNG
To investigate the anti-inflammatory properties of ripasudil, a Rho kinase (ROCK) inhibitor, using endotoxin-induced uveitis (EIU) in rats. Endotoxin-induced uveitis was induced by footpad injection of lipopolysaccharide (LPS). Ripasudil was administered intraperitoneally 1 hour before and after LPS injection. The aqueous humor was collected 24 hours after injection, and the infiltrating cells, protein concentration, and levels of monocyte chemotactic protein-1 (MCP-1) were determined. Infiltrating cells in the iris ciliary body (ICB) and adherent leukocytes in retinal vessels were evaluated. The mRNA levels of IL-1β, IL-6, TNF-α, and MCP-1 in the retina and ICB were determined. A mouse macrophage cell line, RAW264.7, was stimulated with LPS in the presence or absence of ripasudil, and the expression of MCP-1 and nuclear translocation of nuclear factor (NF)-κB was analyzed. Ripasudil significantly reduced infiltrating cells and protein exudation in the aqueous humor, as well as the number of infiltrating cells in the ICB and adherent leukocytes in retinal vessels in EIU. Additionally, the protein level of MCP-1 in the aqueous humor and mRNA levels of IL-1β, IL-6, TNF-α, MCP-1, and intercellular adhesion molecule-1 in the ICB and retina were suppressed by ripasudil. The production of MCP-1 and nuclear translocation of NF-κB in RAW264.7 cells were also suppressed by ripasudil. The Rho/ROCK pathway plays a role in adhesion molecule expression and inflammatory cell infiltration in EIU, and ripasudil is a potent anti-inflammatory agent against ocular inflammatory diseases, including acute uveitis and possibly uveitic glaucoma.