Merck
  • Inhibitors of poly(ADP-ribose) polymerase suppress nuclear fragmentation and apoptotic-body formation during apoptosis in HL-60 cells.
Inhibitors of poly(ADP-ribose) polymerase suppress nuclear fragmentation and apoptotic-body formation during apoptosis in HL-60 cells.
FEBS letters (1997-08-11)
D Shiokawa, H Maruta, S Tanuma
ZUSAMMENFASSUNG
The effects of 3-aminobenzamide (3ABm) and benzamide (BAm), known specific inhibitors of poly(ADP-ribose) polymerase (PARP), on actinomycin D (Act D)-induced apoptosis in HL-60 cells were examined. These inhibitors had no appreciable effect on apoptotic DNA fragmentation, chromatin condensation or PARP restriction cleavage, but clearly inhibited morphological changes, especially nuclear fragmentation and apoptotic-body formation, in a dose-dependent manner. These results suggest that the synthesis of ADP-ribose polymers is not essential for the progression of apoptotic DNA fragmentation and chromatin condensation, but is required in the processes leading to nuclear fragmentation and the subsequent apoptotic-body formation during apoptosis in HL-60 cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-Aminobenzoesäure, 98%