Merck
Adrenal C11-oxy C
The Journal of steroid biochemistry and molecular biology (2017-08-05)
Lise Barnard, Rachelle Gent, Desmaré van Rooyen, Amanda C Swart
ZUSAMMENFASSUNG
21-Hydroxylase deficiency presents with increased levels of cytochrome P450 21-hydroxylase substrates, progesterone and 17α-hydroxyprogesterone, which have been implicated in the production of androgens via the backdoor pathway. This study shows the biosynthesis of C11-oxy C
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
4-Androsten-3,17-dion, VETRANAL®, analytical standard