Merck
  • Structure-Property Relationship Study of Donor and Acceptor 2,6-Disubstituted BODIPY Derivatives for High Performance Dye-Sensitized Solar Cells.
Structure-Property Relationship Study of Donor and Acceptor 2,6-Disubstituted BODIPY Derivatives for High Performance Dye-Sensitized Solar Cells.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-08-02)
Shih-Chieh Yeh, Li-Jing Wang, Hong-Ming Yang, Yu-Huei Dai, Chao-Wen Lin, Chin-Ti Chen, Ru-Jong Jeng
ZUSAMMENFASSUNG
Seven donor and acceptor 2,6-disubstituted 4,4-difluoro-4-bora-3a,4a-diaza-s-indacene (BODIPY) dyes have been synthesized and characterized. Including MPBTCA, which is a known compound, the seven BODIPY dyes have been characterized by varied physical methods, such as UV/Visible absorption spectroscopy, low energy photo-electron spectroscopy (AC-2), and HOMO-LUMO DFT/TDDFT calculation. All seven BODIPY dyes have absorption λ
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-tert-Butylpyridin, 98%
Sigma-Aldrich
1-Butyl-3-methyl-imidazolium-iodid, 99%