Merck
  • MicroRNA-34a functions as a tumor suppressor by directly targeting oncogenic PLCE1 in Kazakh esophageal squamous cell carcinoma.
MicroRNA-34a functions as a tumor suppressor by directly targeting oncogenic PLCE1 in Kazakh esophageal squamous cell carcinoma.
Oncotarget (2017-12-02)
Xiao-Bin Cui, Hao Peng, Ran-Ran Li, Jian-Qin Mu, Lan Yang, Na Li, Chun-Xia Liu, Jian-Ming Hu, Shu-Gang Li, Yutao Wei, Laibo-Yin, Hong Zhou, Feng Li, Yun-Zhao Chen
ZUSAMMENFASSUNG
Esophageal squamous cell carcinoma (ESCC) is one of the frequent malignant tumors with poor prognosis worldwide. Identifying the prognostic biomarkers and potential mechanisms of such tumors has attracted increasing interest in esophageal cancer biology. Our previous study showed that phospholipase C elipson 1 (PLCE1) expression is up-regulated and associated with disease progression in esophageal carcinoma. MicroRNAs (miRNAs) play vital roles in regulating its target gene expression. However, studies on miRNA-regulated PLCE1 expression and its cellular function are still very few. We found that miR-34a is significantly expressed lower in ESCC tissues. We further showed that PLCE1 is a direct functional target gene of miR-34a, and the functional roles of miR-34a in ESCC cell lines
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human PLCE1