Merck
Clone wars: asexual reproduction dominates in the invasive range of
PeerJ (2017-10-12)
Katia Cristina Cruz Capel, Robert J Toonen, Caio T C C Rachid, Joel C Creed, Marcelo V Kitahara, Zac Forsman, Carla Zilberberg
ZUSAMMENFASSUNG
Although the invasive azooxanthellate corals
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
6-Carboxyfluorescein, ≥96% (HPLC)