- Planar and 3D fibrous polyaniline-based materials for memristive elements.
Planar and 3D fibrous polyaniline-based materials for memristive elements.
Soft matter (2017-10-05)
Yulia N Malakhova, Alexei N Korovin, Dmitry A Lapkin, Sergey N Malakhov, Valentina V Shcherban, Eugene B Pichkur, Sergey N Yakunin, Vyacheslav A Demin, Sergey N Chvalun, Victor Erokhin
PMID28976529
ZUSAMMENFASSUNG
The memristive elements constructed using polymers - polyaniline (PANI) and polyethyleneoxide (PEO) - could be assembled on planar thin films or on 3D fibrous materials. Planar conductive PANI-based materials were made using the Langmuir-Schaefer (LS) method, and the 3D materials - using the electrospinning method which is a scalable technique. We have analyzed the influence of PANI molar mass, natures of solvent and subphase on the crystalline structure, thickness and conductivity of planar LS films, and the influence of PANI molar mass and the PANI-PEO ratio on the morphological and structural characteristics of 3D fibrous materials.
MATERIALIEN