Merck
Induction of DISE in ovarian cancer cells
Oncotarget (2017-11-22)
Andrea E Murmann, Kaylin M McMahon, Ashley Haluck-Kangas, Nandini Ravindran, Monal Patel, Calvin Y Law, Sonia Brockway, Jian-Jun Wei, C Shad Thaxton, Marcus E Peter
ZUSAMMENFASSUNG
The death receptor CD95/Fas can be activated by immune cells to kill cancer cells. shRNAs and siRNAs derived from CD95 or CD95 ligand (CD95L) are highly toxic to most cancer cells. We recently found that these sh/siRNAs kill cancer cells in the absence of the target by targeting the 3'UTRs of critical survival genes through canonical RNAi. We have named this unique form of off-target effect DISE (for death induced by survival gene elimination). DISE preferentially kills transformed cells and cancer stem cells. We demonstrate that DISE induction occurs in cancer cells
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Mammalian shRNA Control Transduction Particles, Targets no known mammalian genes