- Production, purification, and assay of streptolysin O.
Production, purification, and assay of streptolysin O.
Methods in enzymology (1988-01-01)
J E Alouf, C Geoffroy
PMID3231118
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Streptolysin O aus Streptococcus pyogenes, ≥1,000,000 units/mg protein, recombinant, lyophilized powder, expressed in E. coli