Merck
  • Discovery of naturally occurring ESR1 mutations in breast cancer cell lines modelling endocrine resistance.
Discovery of naturally occurring ESR1 mutations in breast cancer cell lines modelling endocrine resistance.
Nature communications (2017-12-02)
Lesley-Ann Martin, Ricardo Ribas, Nikiana Simigdala, Eugene Schuster, Sunil Pancholi, Tencho Tenev, Pascal Gellert, Laki Buluwela, Alison Harrod, Allan Thornhill, Joanna Nikitorowicz-Buniak, Amandeep Bhamra, Marc-Olivier Turgeon, George Poulogiannis, Qiong Gao, Vera Martins, Margaret Hills, Isaac Garcia-Murillas, Charlotte Fribbens, Neill Patani, Zheqi Li, Matthew J Sikora, Nicholas Turner, Wilbert Zwart, Steffi Oesterreich, Jason Carroll, Simak Ali, Mitch Dowsett
ZUSAMMENFASSUNG
Resistance to endocrine therapy remains a major clinical problem in breast cancer. Genetic studies highlight the potential role of estrogen receptor-α (ESR1) mutations, which show increased prevalence in the metastatic, endocrine-resistant setting. No naturally occurring ESR1 mutations have been reported in in vitro models of BC either before or after the acquisition of endocrine resistance making functional consequences difficult to study. We report the first discovery of naturally occurring ESR1