Merck
  • Sophoridine from Sophora Flower Attenuates Ovariectomy Induced Osteoporosis through the RANKL-ERK-NFAT Pathway.
Sophoridine from Sophora Flower Attenuates Ovariectomy Induced Osteoporosis through the RANKL-ERK-NFAT Pathway.
Journal of agricultural and food chemistry (2017-10-24)
Xiaoying Zhao, Lijuan Mei, Jinjin Pei, Zenggen Liu, Yun Shao, Yanduo Tao, Xiaoling Zhang, Lei Jiang
ZUSAMMENFASSUNG
An imbalance in osteogenesis and osteoclastogenesis is a crucial pathological factor in the development of osteoporosis. Osteoclasts (OCs) play a pivotal role in osteoporosis, whose new therapy exploration has been focused on the suppression of OC formation. Sophoridine is found from the Chinese traditional food sophora flower to exhibit anti-osteoporosis capacity by screening. This study is focused on its anti-osteoporosis mechanism evaluation. The anti-osteoporosis effect of sophoridine, (15 mg kg
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)