Merck
  • Nanolayered hybrid mediates synergistic co-delivery of ligand and ligation activator for inducing stem cell differentiation and tissue healing.
Nanolayered hybrid mediates synergistic co-delivery of ligand and ligation activator for inducing stem cell differentiation and tissue healing.
Biomaterials (2017-10-11)
Heemin Kang, Minkyu Kim, Qian Feng, Sien Lin, Kongchang Wei, Rui Li, Chan Ju Choi, Tae-Hyun Kim, Gang Li, Jae-Min Oh, Liming Bian
ZUSAMMENFASSUNG
Cellular behaviors, such as differentiation, are regulated by complex ligation processes involving cell surface receptors, which can be activated by various divalent metal cations. The design of nanoparticle for co-delivery of ligand and ligation activator can offer a novel strategy to synergistically stimulate ligation processes in vivo. Here, we present a novel layered double hydroxide (LDH)-based nanohybrid (MgFe-Ado-LDH), composed of layered MgFe hydroxide nanocarriers sandwiching the adenosine cargo molecule, maintained through an electrostatic balance, to co-deliver the adenosine (Ado) ligand from the interlayer spacing and the Mg
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acrylsäure N-Hydroxysuccinimid-Ester, ≥90% (HPLC)