Merck
  • Immunometabolic profiling of T cells from patients with relapsing-remitting multiple sclerosis reveals an impairment in glycolysis and mitochondrial respiration.
Immunometabolic profiling of T cells from patients with relapsing-remitting multiple sclerosis reveals an impairment in glycolysis and mitochondrial respiration.
Metabolism: clinical and experimental (2017-11-15)
Claudia La Rocca, Fortunata Carbone, Veronica De Rosa, Alessandra Colamatteo, Mario Galgani, Francesco Perna, Roberta Lanzillo, Vincenzo Brescia Morra, Giuseppe Orefice, Ilaria Cerillo, Ciro Florio, Giorgia Teresa Maniscalco, Marco Salvetti, Diego Centonze, Antonio Uccelli, Salvatore Longobardi, Andrea Visconti, Giuseppe Matarese
ZUSAMMENFASSUNG
Metabolic reprogramming is shaped to support specific cell functions since cellular metabolism controls the final outcome of immune response. Multiple sclerosis (MS) is an autoimmune disease resulting from loss of immune tolerance against central nervous system (CNS) myelin. Metabolic alterations of T cells occurring during MS are not yet well understood and their studies could have relevance in the comprehension of the pathogenetic events leading to loss of immune tolerance to self and to develop novel therapeutic strategies aimed at limiting MS progression. In this report, we observed that extracellular acidification rate (ECAR) and oxygen consumption rate (OCR), indicators of glycolysis and oxidative phosphorylation, respectively, were impaired during T cell activation in naïve-to-treatment relapsing remitting (RR)MS patients when compared with healthy controls. These results were also corroborated at biochemical level by a reduced expression of the glycolitic enzymes aldolase, enolase 1, hexokinase I, and by reduction of Krebs cycle enzymes dihydrolipoamide-S-acetyl transferase (DLAT) and dihydrolipoamide-S-succinyl transferase (DLST). Treatment of RRMS patients with interferon beta-1a (IFN beta-1a) was able to restore T cell glycolysis and mitochondrial respiration as well as the amount of the metabolic enzymes to a level comparable to that of healthy controls. These changes associated with an up-regulation of the glucose transporter-1 (GLUT-1), a key element in intracellular transport of glucose. Our data suggest that T cells from RRMS patients display a reduced engagement of glycolysis and mitochondrial respiration, reversible upon IFN beta-1a treatment, thus suggesting an involvement of an altered metabolism in the pathogenesis of MS.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phosphatasehemmer-Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Carbonylcyanid-4-(trifluormethoxy)phenylhydrazon, ≥98% (HPLC), powder
Sigma-Aldrich
SIGMAFAST Proteaseinhibitor-Tabletten, For General Use