Merck
Rods progressively escape saturation to drive visual responses in daylight conditions.
Nature communications (2017-11-29)
Alexandra Tikidji-Hamburyan, Katja Reinhard, Riccardo Storchi, Johannes Dietter, Hartwig Seitter, Katherine E Davis, Saad Idrees, Marion Mutter, Lauren Walmsley, Robert A Bedford, Marius Ueffing, Petri Ala-Laurila, Timothy M Brown, Robert J Lucas, Thomas A Münch
ZUSAMMENFASSUNG
Rod and cone photoreceptors support vision across large light intensity ranges. Rods, active under dim illumination, are thought to saturate at higher (photopic) irradiances. The extent of rod saturation is not well defined; some studies report rod activity well into the photopic range. Using electrophysiological recordings from retina and dorsal lateral geniculate nucleus of cone-deficient and visually intact mice, we describe stimulus and physiological factors that influence photopic rod-driven responses. We find that rod contrast sensitivity is initially strongly reduced at high irradiances, but progressively recovers to allow responses to moderate contrast stimuli. Surprisingly, rods recover faster at higher light levels. A model of rod phototransduction suggests that phototransduction gain adjustments and bleaching adaptation underlie rod recovery. Consistently, exogenous chromophore reduces rod responses at bright background. Thus, bleaching adaptation renders mouse rods responsive to modest contrast at any irradiance. Paradoxically, raising irradiance across the photopic range increases the robustness of rod responses.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
9-cis-Retinal, vitamin A analog
Sigma-Aldrich
L-(+)-2-Amino-4-phosphonobutyric acid, optical purity optical purity: ≥95% (HPLC, Marfey′s reagent)