Merck
  • Simultaneous targeted inhibition of Sox2-Oct4 transcription factors using decoy oligodeoxynucleotides to repress stemness properties in mouse embryonic stem cells.
Simultaneous targeted inhibition of Sox2-Oct4 transcription factors using decoy oligodeoxynucleotides to repress stemness properties in mouse embryonic stem cells.
Cell biology international (2017-08-24)
Behrooz Johari, Jamil Zargan
ZUSAMMENFASSUNG
Transcriptional master regulators like Sox2 and Oct4, which are expressed in various human tumors, have been shown to cause tumor growth promotion as well as epithelial dysplasia by means of interfering with progenitor cell differentiation. In order to investigate the potential of Sox2-Oct4 transcription factor decoy (TFD) strategy for differentiation therapy, mouse embryonic stem cells (mESCs) were used in this study as a model of cancer stem cells (CSCs). Sox2-Oct4 complex decoy ODNs (cd-ODNs) were designed according to their elements in the promoter region of Sox2 gene. DNA-protein interactions between decoy ODNs and their corresponding proteins were examined by electrophoretic mobility shift assay (EMSA). Then, decoy and scrambled ODNs were transfected into mESCs with lipofectamine under 2 inhibitors (2i) conditions. Fluorescence and confocal microscopy, cell viability, cell cycle and apoptosis analysis, alkaline phosphatase, embryoid body formation assay, and real-time PCR were used to conduct further investigations. EMSA data showed that Sox2-Oct4 decoy ODNs bound specifically to their recombinant proteins. The results revealed that the synthesized complex decoy can concomitantly target Sox2 and Oct4, which subsequently represses the stemness properties of mESCs compared to controls through decreasing cell viability, arresting cell cycle in G
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Sigma-Aldrich
2-Phenylindol, technical grade, 95%