Merck
  • A Modified Carbapenem Inactivation Method, CIMTris, for Carbapenemase Production in Acinetobacter and Pseudomonas Species.
A Modified Carbapenem Inactivation Method, CIMTris, for Carbapenemase Production in Acinetobacter and Pseudomonas Species.
Journal of clinical microbiology (2017-09-29)
Kohei Uechi, Tatsuya Tada, Kayo Shimada, Kyoko Kuwahara-Arai, Momoko Arakaki, Takaaki Tome, Isamu Nakasone, Shiro Maeda, Teruo Kirikae, Jiro Fujita
ZUSAMMENFASSUNG
The carbapenem inactivation method (CIM) and modified CIM (mCIM) are simple and economical phenotypic screening methods for detecting carbapenemase production in Gram-negative bacteria. Although the mCIM has been recommended by the Clinical and Laboratory Standards Institute, both the CIM and mCIM have limitations. This study describes another modified CIM, called CIMTris, in which carbapenemase was extracted from bacteria with 0.5 M Tris-HCl (pH 7.6) buffer. The ability of the CIMTris to detect carbapenemase production was examined in