Merck
  • Influence of development, postharvest handling, and storage conditions on the carbohydrate components of sweetpotato (
Influence of development, postharvest handling, and storage conditions on the carbohydrate components of sweetpotato (
Food science & nutrition (2017-12-01)
Agnes Nabubuya, Agnes Namutebi, Yusuf Byaruhanga, Judith Narvhus, Trude Wicklund
ZUSAMMENFASSUNG
Changes in total starch and reducing sugar content in five sweetpotato varieties were investigated weekly during root development and following subjection of the roots to different postharvest handling and storage conditions. Freshly harvested (noncured) roots and cured roots (spread under the sun for 4 days at 29-31°C and 63-65% relative humidity [RH]) were separately stored at ambient conditions (23°C-26°C and 70-80% RH) and in a semiunderground pit (19-21°C and 90-95% RH). Changes in pasting properties of flour from sweetpotato roots during storage were analyzed at 14-day intervals. Significant varietal differences (
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Maltose -Lösung, BioReagent, ~20% in H2O, Molecular Biology