Merck
Systematic characterization of the effect of Ag@TiO
Scientific reports (2017-11-18)
Pascal Nbelayim, Go Kawamura, Wai Kian Tan, Hiroyuki Muto, Atsunori Matsuda
ZUSAMMENFASSUNG
The use of plasmonic nanoparticles (NPs) in dye-sensitized solar cells (DSSCs) in an effort to enhance their power conversion efficiencies (PCEs) increases light absorbance of the cells but also affect their electron dynamics. This has contributed to the failure of plasmonic NPs to make the expected high impact of PCE enhancement. Herein, we investigated the wide range effects of plasmonic NPs on the performance of DSSCs, using extended characterization and a systematic approach. We prepared DSSCs using Ag@TiO
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-tert-Butylpyridin, 98%