Merck
  • Fabrication of engineered biochar from paper mill sludge and its application into removal of arsenic and cadmium in acidic water.
Fabrication of engineered biochar from paper mill sludge and its application into removal of arsenic and cadmium in acidic water.
Bioresource technology (2017-07-18)
Kwangsuk Yoon, Dong-Wan Cho, Daniel C W Tsang, Nanthi Bolan, Jörg Rinklebe, Hocheol Song
ZUSAMMENFASSUNG
An engineered biochar was fabricated via paper mill sludge pyrolysis under CO
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Calcium Standard für AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM