Merck
Extraction, structure, and emulsifying properties of pectin from potato pulp.
Food chemistry (2017-11-10)
Jin-Shu Yang, Tai-Hua Mu, Meng-Mei Ma
ZUSAMMENFASSUNG
Effects of HCl, H
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG